Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF39 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155282 Supplier Novus Biologicals Supplier No. NBP155282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF39 Polyclonal specifically detects RNF39 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RNF39
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. A2BEK3
Gene Alias HZF, HZFWLTP (long-term potentiation) induced RING finger protein, LIRFHZFw1, Protein HZFw, ring finger protein 39
Gene Symbols RNF39
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RNF39(ring finger protein 39) The peptide sequence was selected from the N terminal of RNF39. Peptide sequence EGRKAAKVNAGVGEKGIYTASSRGGPPSARSKAVTVVAEGAASRSWLSMD.
Molecular Weight of Antigen 39 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80352
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.