Learn More
RNF39 Mouse anti-Human, Clone: OTI4D3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505914

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
RNF39 | |
Monoclonal | |
Unconjugated | |
RNF39 | |
HZF, HZFW, LIRF | |
Mouse | |
Affinity Chromatography | |
RUO | |
80352 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4D3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H2S5 | |
RNF39 | |
Full length human recombit protein of human RNF39 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.