Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF39 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561431
Description
Immunogen sequence: SLRSNVRLAV EVRISRELRE KLAEPGARAG RRRGGRIPTM GCLDLPGEDM RKTWRRFEVP TSKSSNSEDD LPEDYPVVKK MLHRLTADLT LDP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
RNF39 | |
Polyclonal | |
Unconjugated | |
Rnf39 | |
BC038661; DADB-60O11.8; Gm1786; Gm1787; Gm320; HZF; HZFW; Hzfw1; HZFw1 protein; Lirf; LTP (long-term potentiation) induced RING finger protein; LTP-induced RING finger protein; Protein HZFw; RING finger protein 39; RNF39 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80352 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2S5 | |
Rnf39 | |
Recombinant protein corresponding to Human RNF39. Recombinant protein control fragment (Product #RP-99611). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction