Learn More
Invitrogen™ RNF39 Polyclonal Antibody
Description
RNF39 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
Specifications
| Antigen | RNF39 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | Rnf39 |
| Gene Accession No. | Q920M2, Q9H2S5 |
| Gene Alias | BC038661; DADB-60O11.8; Gm1786; Gm1787; Gm320; HZF; HZFW; Hzfw1; HZFw1 protein; Lirf; LTP (long-term potentiation) induced RING finger protein; LTP-induced RING finger protein; Protein HZFw; RING finger protein 39; RNF39 |
| Gene Symbols | Rnf39 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.