Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF4 Monoclonal Antibody (OTI9F10)
GREENER_CHOICE

Catalog No. PIMA527424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527424 Supplier Invitrogen™ Supplier No. MA527424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNF4 Monoclonal Antibody for Western Blot

The protein encoded by this gene contains a RING finger motif and acts as a transcription regulator. This protein has been shown to interact with, and inhibit the activity of, TRPS1, a transcription suppressor of GATA-mediated transcription. Transcription repressor ZNF278/PATZ is found to interact with this protein, and thus reduce the enhancement of androgen receptor-dependent transcription mediated by this protein. Studies of the mouse and rat counterparts suggested a role of this protein in spermatogenesis. A pseudogene of this gene is found on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF4
Applications Western Blot
Classification Monoclonal
Clone OTI9F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNF4
Gene Accession No. O88846, P78317, Q9QZS2
Gene Alias AU018689; E3 ubiquitin ligase RNF4; E3 ubiquitin-protein ligase RNF4; Gtrgeo8; Protein SNURF; RES4-26; RING finger protein 4; RING-type E3 ubiquitin transferase RNF4; RNF4; SLX5; small nuclear RING finger protein; SNURF
Gene Symbols RNF4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-190 of RNF4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19822, 29274, 6047
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.