Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF40 Antibody (SR1131), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | RNF40 |
---|---|
Clone | SR1131 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
RNF40 Monoclonal antibody specifically detects RNF40 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
RNF40 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
95 kDa retinoblastoma-associated protein, BRE1-B, BRE1BMGC13051, EC 6.3.2,95 kDa retinoblastoma protein binding protein, EC 6.3.2.-, KIAA0661BRE1 E3 ubiquitin ligase homolog B, Rb-associated protein, RBP95E3 ubiquitin-protein ligase BRE1B, ring finger protein 40DKFZp686K191, STARING | |
A synthesized peptide derived from human RNF40 (Uniprot #: O75150) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1131 | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Chromatin Research, Vision, Zinc Finger | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
9810 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title