Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF40 Recombinant Rabbit Monoclonal Antibody (8I7R5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535816
Description
Immunogen sequence: RELGRENRRL QDLATQLQEK HHRISLEYSE LQDKVTSAET KVLEMETTVE DLQWDIEKLR KREQKLNKHL AEALEQLNSG YYVSGSSSGF QGGQITLSMQ KFEMLNAELE ENQELANSRM AELEKLQAEL QGAVRTNERL KVALRSLPEE VVRETGEYRM LQAQFSLLYN ESLQVKTQLD EARGLLLATK NSHLRHIEHM E.
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery.
Specifications
RNF40 | |
Recombinant Monoclonal | |
1.65 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75150, Q3U319 | |
RNF40 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 231-431 of human RNF40 (O75150). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
8I7R5 | |
Unconjugated | |
RNF40 | |
95 kDa retinoblastoma protein binding protein; 95 kDa retinoblastoma-associated protein; AI848422; BRE1 E3 ubiquitin ligase homolog B; Bre1b; BRE1-B; DKFZp686K191; E3 ubiquitin-protein ligase BRE1B; KIAA0661; MGC13051; mKIAA0661; protein staring; Rb-associated protein; RBP95; RING finger protein 40; ring finger protein 40, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1B; Rnf40; STARING; staring protein; syntaxin-1-interacting RING finger protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
233900, 9810 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction