Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592980 Supplier Invitrogen™ Supplier No. PA592980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSGPGNKRAA GDGGSGPPEK KLSREEKTTT TLIEPIRLGG ISSTEEMDLK VLQFKNKKLA ERLEQRQACE DELRERIEKL EKRQATDDAT LLIVNRYWAQ LDETVEALLR CHESQGELSS APEAPGTQEG PTCDGTPLPE PGTSELRDPL LMQLRPPLSE PALAFVVALG ASSSEEVELE LQGRMEFSKA AVSRVVEASD RLQRRVEELC QRVYSRGDSE PLSEAAQAHT RELGRENRRL QDLATQLQEK HHRISLEYSE LQDKVTSAET KVLEMETTVE DLQWDIEKLR KREQKLNKHL; Positive Samples: MCF7, HepG2, U-937; Cellular Location: Nucleus.

The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF40
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNF40
Gene Accession No. O75150
Gene Alias 95 kDa retinoblastoma protein binding protein; 95 kDa retinoblastoma-associated protein; AI848422; BRE1 E3 ubiquitin ligase homolog B; Bre1b; BRE1-B; DKFZp686K191; E3 ubiquitin-protein ligase BRE1B; KIAA0661; MGC13051; mKIAA0661; protein staring; Rb-associated protein; RBP95; RING finger protein 40; ring finger protein 40, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1B; Rnf40; STARING; staring protein; syntaxin-1-interacting RING finger protein
Gene Symbols RNF40
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RNF40 (NP_0555861).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9810
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.