Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF41 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RNF41 |
---|---|
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RNF41 Polyclonal specifically detects RNF41 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RNF41 | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
E3 ubiquitin-protein ligase NRDP1, EC 6.3.2, EC 6.3.2.-, FLRF, MGC45228, NRDP1, ring finger protein 41neuregulin receptor degradation protein-1, SBBI03 | |
RNF41 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
Q9H4P4 | |
10193 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DAVLQAVIKRSLVESGCPASIVNELIENAHERSWPQGLATLETRQMNRRYYENYVAKRIPGKQAVVVMACENQHMGDDMVQEPGLVMIFAHGVEEI | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title