Learn More
RNF41 Mouse anti-Human, Clone: OTI2C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811158

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. The specific function of this protein has not yet been determined. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported.Specifications
RNF41 | |
Monoclonal | |
Unconjugated | |
RNF41 | |
FLRF, NRDP1, SBBI03 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10193 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H4P4 | |
RNF41 | |
Full length human recombit protein of human RNF41 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.