Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553495 Supplier Invitrogen™ Supplier No. PA553495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAVLQAVIKR SLVESGCPAS IVNELIENAH ERSWPQGLAT LETRQMNRRY YENYVAKRIP GKQAVVVMAC ENQHMGDDMV QEPGLVMIFA HGVEEI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. Nrdp1, also known as RNF41 (RING finger protein 41), SBBI03 or FLRF, is a 317 amino acid protein that contains one RING-type zinc finger and one SIAH-type zinc finger. Expressed in testis, ovary and prostate, Nrdp1 functions as an E3 ubiquitin-protein ligase that, characteristic of E3 ligase proteins, accepts ubiquitin (in the form of a thioester) from an E2 ubiquitin-conjugating enzyme and transfers that ubiquitin residue to substrates targeted for degradation. Specifically, Nrdp1 interacts with ErbB-3 and UBPY, thereby targeting them for proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF41
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF41
Gene Accession No. Q9H4P4
Gene Alias 2210404G21Rik; 4930511A05Rik; 4933415P08Rik; D10Ertd722e; E3 ubiquitin-protein ligase NRDP1; fetal liver ring finger; FLRF; neuregulin receptor degradation protein-1; Nrdp1; RING finger protein 41; ring finger protein 41, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase NRDP1; Rnf41; SBBI03
Gene Symbols RNF41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF41. Recombinant protein control fragment (Product #RP-91674).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.