Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF43 Antibody (077), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290325AF700
Description
RNF43 Monoclonal specifically detects RNF43 in Human samples. It is validated for ELISA.Specifications
RNF43 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
77 | |
ELISA | |
DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human RNF43 (NP_060233.3; Met1-Tyr197). | |
0.1 mL | |
Cancer | |
54894 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction