Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF43 Antibody (923227R) [DyLight 594], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7964M
Description
RNF43 Monoclonal antibody specifically detects RNF43 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
RNF43 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
923227R | |
Flow Cytometry, CyTOF-ready | |
DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC | |
Human embryonic kidney cell line HEK293-derived recombinant human RNF43, Glu43-Val199, Accession # Q68DV7 | |
0.1 mL | |
Cancer | |
54894 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction