Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF43 Monoclonal Antibody (06)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530820
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RNF43.
E3 ubiquitin-protein ligase that acts as a negative regulator of the Wnt signaling pathway by mediating the ubiquitination, endocytosis and subsequent degradation of Wnt receptor complex components Frizzled. Acts on both canonical and non-canonical Wnt signaling pathway. Acts as a tumor suppressor in the intestinal stem cell zone by inhibiting the Wnt signaling pathway, thereby resticting the size of the intestinal stem cell zone.
Specifications
RNF43 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q68DV7 | |
RNF43 | |
Recombinant Human RNF43 Protein (Met1-Tyr197). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
06 | |
Unconjugated | |
RNF43 | |
4732452J19Rik; E3 ubiquitin-protein ligase RNF43; RGD1305204; RING finger protein 43; RING-type E3 ubiquitin transferase RNF43; RNF124; Rnf43; URCC | |
Mouse | |
Protein A | |
RUO | |
54894 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction