Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552512 Supplier Invitrogen™ Supplier No. PA552512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVDPWLHQHR TCPLCMFNIT EGDSFSQSLG PSRSYQEPGR RLHLIRQHPG HAHYHLPAAY LLGPSRSAVA RPPRPGPFLP SQEPGMGPRH HRFPRAAHPR APGEQQRLAG AQHPYAQGWG LSHLQSTSQH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

E3 ubiquitin-protein ligase that acts as a negative regulator of the Wnt signaling pathway by mediating the ubiquitination, endocytosis and subsequent degradation of Wnt receptor complex components Frizzled. Acts on both canonical and non-canonical Wnt signaling pathway. Acts as a tumor suppressor in the intestinal stem cell zone by inhibiting the Wnt signaling pathway, thereby resticting the size of the intestinal stem cell zone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF43
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF43
Gene Accession No. Q68DV7
Gene Alias 4732452J19Rik; E3 ubiquitin-protein ligase RNF43; RGD1305204; RING finger protein 43; RING-type E3 ubiquitin transferase RNF43; RNF124; Rnf43; URCC
Gene Symbols RNF43
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF43. Recombinant protein control fragment (Product #RP-89654).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.