Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    RNF43 Rabbit anti-Human, Clone: 77, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290325100UL
Description
RNF43 Monoclonal specifically detects RNF43 in Human samples. It is validated for ELISA.Specifications
| RNF43 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 54894 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG | 
| ELISA | |
| 77 | |
| ELISA 1:5000-1:10000 | |
| DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC | |
| Recombinant Human RNF43 Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            