Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF44 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | RNF44 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RNF44 Polyclonal specifically detects RNF44 in Human samples. It is validated for Western Blot.Specifications
RNF44 | |
Polyclonal | |
Rabbit | |
Q7L0R7 | |
22838 | |
Synthetic peptides corresponding to RNF44(ring finger protein 44) The peptide sequence was selected from the N terminal of RNF44. Peptide sequence LSYTVTTVTTQGFPLPTGQHIPGCSAQQLPACSVMFSGQHYPLCCLPPPL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
KIAA1100, ring finger protein 44 | |
RNF44 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title