Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF5 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204014V
Description
RNF5 Polyclonal antibody specifically detects RNF5 in Human samples. It is validated for Western BlotSpecifications
RNF5 | |
Polyclonal | |
Western Blot | |
EC 6.3.2.-, G16, HsRma1, NG2, Protein G16, ring finger protein 5Ram1 homolog, RING5hsRma1, RMA1E3 ubiquitin-protein ligase RNF5 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Ring Finger Protein 5 using the numbering given in entry NP_008844.1 (GeneID 6048). | |
0.1 mL | |
Cancer, Cell Biology, Immunology | |
6048 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction