Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528442
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene contains a RING-H2 finger motif. Deletions and mutations in this gene were detected in esophageal squamous cell carcinoma (ESCC), suggesting that this protein may be a potential tumor suppressor. Studies of the mouse counterpart suggested a role of this protein in the transcription regulation that controls germinal differentiation. Multiple alternatively spliced transcript variants encoding the same protein are observed.
Specifications
RNF6 | |
Polyclonal | |
Unconjugated | |
RNF6 | |
1200013I08Rik; 5730419H05Rik; AA537053; DKFZp686P0776; E3 ubiquitin-protein ligase RNF6; ring finger protein (C3H2C3 type) 6; ring finger protein 6; RING-H2 protein RNF-6; RING-type E3 ubiquitin transferase RNF6; RLIM-like protein; RNF6; SPG2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6049 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.86 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y252 | |
RNF6 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human RNF6. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction