Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558746 Supplier Invitrogen™ Supplier No. PA558746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGTVYHNSQR ESRPVQQTTR RSVRRRGRTR VFLEQDRERE RRGTAYTPFS NSRLVSRITV EEGEESSRSS TAVRR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 72%.

The protein encoded by this gene contains a RING-H2 finger motif. Deletions and mutations in this gene were detected in esophageal squamous cell carcinoma (ESCC), suggesting that this protein may be a potential tumor suppressor. Studies of the mouse counterpart suggested a role of this protein in the transcription regulation that controls germinal differentiation. Multiple alternatively spliced transcript variants encoding the same protein are observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF6
Gene Accession No. Q9Y252
Gene Alias 1200013I08Rik; 5730419H05Rik; AA537053; DKFZp686P0776; E3 ubiquitin-protein ligase RNF6; ring finger protein (C3H2C3 type) 6; ring finger protein 6; RING-H2 protein RNF-6; RING-type E3 ubiquitin transferase RNF6; RLIM-like protein; RNF6; SPG2
Gene Symbols RNF6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF6. Recombinant protein control fragment (Product #RP-102869).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6049
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.