Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568868 Supplier Invitrogen™ Supplier No. PA568868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 93%.

The protein encoded by this gene contains a RING-H2 finger motif. Deletions and mutations in this gene were detected in esophageal squamous cell carcinoma (ESCC), suggesting that this protein may be a potential tumor suppressor. Studies of the mouse counterpart suggested a role of this protein in the transcription regulation that controls germinal differentiation. Multiple alternatively spliced transcript variants encoding the same protein are observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF6
Gene Accession No. Q9DBU5
Gene Alias 1200013I08Rik; 5730419H05Rik; AA537053; DKFZp686P0776; E3 ubiquitin-protein ligase RNF6; ring finger protein (C3H2C3 type) 6; ring finger protein 6; RING-H2 protein RNF-6; RING-type E3 ubiquitin transferase RNF6; RLIM-like protein; RNF6; SPG2
Gene Symbols RNF6
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse RNF6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74132
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.