Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557906
Description
Immunogen sequence: SLKKWNAVAM WSWDVECDTC AICRVQVMDA CLRCQAENKQ EDCVVVWGEC NHSFHNCCMS LWVKQNNRCP LCQQDWVVQR IGK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is a highly conserved ring finger protein. It is an essential subunit of SKP1-cullin/CDC53-F box protein ubiquitin ligases, which are a part of the protein degradation machinery important for cell cycle progression and signal transduction. This protein interacts with, and is a substrate of, casein kinase II (CSNK2A1/CKII). The phosphorylation of this protein by CSNK2A1 has been shown to promote the degradation of IkappaBalpha (CHUK/IKK-alpha/IKBKA) and p27Kip1(CDKN1B). Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
RNF7 | |
Polyclonal | |
Unconjugated | |
RNF7 | |
CKBBP1; CKII beta-binding protein 1; OTTHUMP00000180645; rbx2; Regulator of cullins 2; ring finger protein 7; RING-box protein 2; RNF7; ROC2; SAG; Sensitive to apoptosis gene protein; sensitive to apoptosis, zinc RING finger protein SAG, regulator of cullins 2; zinc RING finger protein SAG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBF6 | |
RNF7 | |
Recombinant protein corresponding to Human RNF7. Recombinant protein control fragment (Product #RP-96284). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction