Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550872
Description
The antibody detects endogenous levels of total RNF7 protein.
The protein encoded by this gene is a highly conserved ring finger protein. It is an essential subunit of SKP1-cullin/CDC53-F box protein ubiquitin ligases, which are a part of the protein degradation machinery important for cell cycle progression and signal transduction. This protein interacts with, and is a substrate of, casein kinase II (CSNK2A1/CKII). The phosphorylation of this protein by CSNK2A1 has been shown to promote the degradation of IkappaBalpha (CHUK/IKK-alpha/IKBKA) and p27Kip1(CDKN1B). Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
RNF7 | |
Polyclonal | |
Unconjugated | |
RNF7 | |
CKBBP1; CKII beta-binding protein 1; OTTHUMP00000180645; rbx2; Regulator of cullins 2; ring finger protein 7; RING-box protein 2; RNF7; ROC2; SAG; Sensitive to apoptosis gene protein; sensitive to apoptosis, zinc RING finger protein SAG, regulator of cullins 2; zinc RING finger protein SAG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19823, 9616 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UBF6, Q9WTZ1 | |
RNF7 | |
Full length fusion protein of Human RNF7. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction