Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF70 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281998
Description
RNF70 Polyclonal specifically detects RNF70 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RNF70 | |
Polyclonal | |
Unconjugated | |
PBS | |
PJA1 | |
RNF70 antibody was raised against a 19 amino acid peptide near the amino terminus of human RNF70. The immunogen is located within amino acids 50 - 100 of RNF70. Amino Acid Squence: QTTKRSRSPFSTTRRSWDD | |
Affinity Purified | |
RUO | |
Primary | |
RNF70 antibody is human, mouse and rat reactive. At least two isoforms of RNF70 are known to exist; this antibody will detect both isoforms. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 0.25-0.5 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
EC 6.3.2, EC 6.3.2.-, FLJ11830, praja 1, praja ring finger 1, PRAJA1, RING finger protein 70, RNF70E3 ubiquitin-protein ligase Praja-1 | |
Rabbit | |
71 kDa | |
0.1 mg | |
Lipid and Metabolism, Zinc Finger | |
64219 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction