Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF8 Antibody (SR2063), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321837100UL
Description
RNF8 Monoclonal antibody specifically detects RNF8 in Human, Rat samples. It is validated for Western BlotSpecifications
RNF8 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot | |
SR2063 | |
Western Blot 1:500-1:2000 | |
C3HC4-type zinc finger protein, EC 6.3.2, EC 6.3.2.-, FLJ12013, KIAA0646UBC13/UEV-interacting ring finger protein, ring finger protein (C3HC4 type) 8, ring finger protein 8E3 ubiquitin-protein ligase RNF8 | |
A synthesized peptide derived from human RNF8 (Uniprot #: O76064) | |
100 μg | |
Stem Cell Markers, Zinc Finger | |
9025 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction