Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530295 Supplier Invitrogen™ Supplier No. PA530295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (82%), Rat (82%), Rhesus Monkey (97%), Chimpanzee (99%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF8
Applications Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RNF8
Gene Accession No. O76064
Gene Alias 3830404E21Rik; ActA binding protein 2; actA-interacting protein 37; AIP37; C3HC4-type zinc finger protein; E3 ubiquitin-protein ligase RNF8; FLJ12013; hRNF8; KIAA0646; LaXp180; OTTHUMP00000039684; ring finger protein (C3HC4 type) 8; ring finger protein 8; ring finger protein 8, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF8; RNF8; UBC13/UEV-interacting ring finger protein
Gene Symbols RNF8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RNF8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9025
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.