Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF98 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988860
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8860 Supplier Bioss Supplier No. BS9155R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.

Specifications

Antigen RNF98
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TRIM36
Gene Accession No. Q9NQ86
Gene Alias Acrosome RBCC protein; BOS_10103; D18Wsu100e; E3 ubiquitin-protein ligase TRIM36; Haprin; LOC100344463; RBCC728; RING finger protein 98; RING-type E3 ubiquitin transferase TRIM36; RNF98; TRIM36; tripartite motif containing 36; tripartite motif protein 36; tripartite motif-containing 36; Tripartite motif-containing protein 36; wu:fj48f01; zgc:153447; zgc:63476; Zinc-binding protein Rbcc728
Gene Symbols TRIM36
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TRIM36/RNF98.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55521
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.