Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNGTT Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297837JF646
Description
RNGTT Polyclonal specifically detects RNGTT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RNGTT | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CAP1AmRNA-capping enzyme, hCAP, HCE1DKFZp686J2031, HCEHCAP1, RNA guanylyltransferase and 5'-phosphatase | |
Produced in rabbits immunized with E. coli-derived Human RNGTT fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
8732 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction