Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNGTT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 124301AP150UL

Description
Bifunctional mRNA-capping enzyme exhibiting RNA 5′-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5′-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5′-diphosphate terminus.Specifications
RNGTT | |
Polyclonal | |
Unconjugated | |
RNGTT | |
CAP1A, GTase, GTP RNA guanylyltransferase, hCAP, HCAP1, HCE, HCE1, mRNA 5 triphosphatase, mRNA capping enzyme, RNGTT, TPase mRNA guanylyltransferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24018, 313131, 8732 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O55236, O60942 | |
RNGTT | |
RNGTT Fusion Protein Ag3105 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction