Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNH1 Monoclonal Antibody (4F3)
GREENER_CHOICE

Catalog No. PIMA549270
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549270 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549270 Supplier Invitrogen™ Supplier No. MA549270
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is different from the related mouse sequence by five amino acids, and from the related rat sequence by four amino acids. Positive Control - WB: human Hela whole cell, human Jurkat whole cell, human 293T whole cell, human placenta tissue. IHC: human mammary cancer tissue, human ovarian cancer tissue, human prostatic cancer tissue. ICC/IF: Hela cell. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNH1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4F3
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene RNH1
Gene Accession No. P13489
Gene Alias AW546468; C80305; EGK_05816; hypothetical protein LOC517087; im:7039360; LOC720791; Placental ribonuclease inhibitor; placental RNase inhibitor; PRI; RAI; Ribonuclease inhibitor; ribonuclease/angiogenin inhibitor 1; RNasIN; RNH; Rnh1; si:ch211-213a13.3; testicular tissue protein Li 164; uncharacterized protein rnh1; wu:fd55g05
Gene Symbols RNH1
Host Species Mouse
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human RNH1 (2-37aa SLDIQSLDIQCEELSDARWAELLPLLQQCQVVRLDD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6050
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.