Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNH1 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
SDP

Catalog No. 502541889
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1889 30 μL
50-167-3060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1889 Supplier OriGene Supplier No. TA501965S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNH1 Monoclonal antibody specifically detects RNH1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RNH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B1
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pecr
Gene Accession No. Q9BY49
Gene Alias AW546468; C80305; EGK_05816; hypothetical protein LOC517087; im:7039360; LOC720791; Placental ribonuclease inhibitor; placental RNase inhibitor; PRI; RAI; Ribonuclease inhibitor; ribonuclease/angiogenin inhibitor 1; RNH; Rnh1; si:ch211-213a13.3; testicular tissue protein Li 164; uncharacterized protein rnh1; wu:fd55g05
Host Species Mouse
Immunogen Full length human recombinant protein of human PECR produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55825
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.