Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNH1 Monoclonal Antibody (OTI4G4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501874S


Description
RNH1 Monoclonal antibody specifically detects RNH1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
RNH1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O60861 | |
Mouse | |
Affinity chromatography | |
RUO | |
8522 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4G4 | |
Unconjugated | |
GAS7 | |
AW546468; C80305; EGK_05816; hypothetical protein LOC517087; im:7039360; LOC720791; Placental ribonuclease inhibitor; placental RNase inhibitor; PRI; RAI; Ribonuclease inhibitor; ribonuclease/angiogenin inhibitor 1; RNH; Rnh1; si:ch211-213a13.3; testicular tissue protein Li 164; uncharacterized protein rnh1; wu:fd55g05 | |
Full length human recombinant protein of human GAS7 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction