Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNH1 Mouse anti-Human, Clone: 4A1C4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | RNH1 |
---|---|
Clone | 4A1C4 |
Concentration | 1.18 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6469
![]() ![]() |
Proteintech Group Inc
660281IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.Specifications
RNH1 | |
1.18 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Placental RNase inhibitor, PRI, RAI, Ribonuclease inhibitor, RNH, RNH1 | |
RNH1 | |
IgG2b | |
Protein A | |
Antibody |
4A1C4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P13489 | |
6050 | |
RNH1 Fusion Protein Ag18093 | |
Primary | |
-20°C | |
RNH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title