Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596272 Supplier Invitrogen™ Supplier No. PA596272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALKLESCGV TSDNCRDLCG IVASKASLRE LALGSNKLGD VGMAELCPGL LHPSSRLRTL WIWECGITAK GCGDLCRVLR AKESLKELSL AGNELGDEGA RLLCETLLEP GCQLESLWVK SCSFTAACCS HFSSVLAQNR FLLELQISNN RLEDAGVREL CQGLGQPGSV LRVLWLADCD VSDSSCSSLA ATLLANHSLR ELDLSNNCLG DAGILQLVES VRQPGCLLEQ LVLYDIYWSE EMEDRLQALE KDKPSLRVIS; Positive Samples: Raji, U-87MG, HT-29, HeLa, Mouse liver, Mouse kidney, Mouse lung, Rat brain, Rat lung; Cellular Location: Cytoplasm.

RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNH1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNH1
Gene Accession No. P13489, P29315, Q91VI7
Gene Alias AW546468; C80305; EGK_05816; hypothetical protein LOC517087; im:7039360; LOC720791; Placental ribonuclease inhibitor; placental RNase inhibitor; PRI; RAI; Ribonuclease inhibitor; ribonuclease/angiogenin inhibitor 1; RNasIN; RNH; Rnh1; si:ch211-213a13.3; testicular tissue protein Li 164; uncharacterized protein rnh1; wu:fd55g05
Gene Symbols RNH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 202-461 of human RNH1 (NP_0029302).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360501, 107702, 6050
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.