Learn More
RNH1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305356AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.Specifications
RNH1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P13489, Q91VI7 | |
Rnh1 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RNH1 | |
placental ribonuclease inhibitor, placental RNase inhibitor, RAI, ribonuclease/angiogenin inhibitor, Ribonuclease/angiogenin inhibitor 1, RNH, testicular tissue protein Li 164 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107702, 6050 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.