Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNMT Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297644JF549
Description
RNMT Polyclonal specifically detects RNMT in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RNMT | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DKFZp686H1252, EC 2.1.1.56, hcm1p, hCMT1, hCMT1c, hMet, KIAA0398, mRNA (guanine-7-)methyltransferase, mRNA (guanine-N(7)-)-methyltransferase, mRNA cap guanine-N7 methyltransferase, mRNA cap methyltransferase, RG7MT1MET, RNA (guanine-7-) methyltransferase | |
Produced in rabbits immunized with E. coli-derived Human RNMT fragment. | |
0.1 mL | |
Phospho Specific | |
8731 | |
Store at 4°C in the dark. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction