Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112213 Supplier Invitrogen™ Supplier No. PA5112213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNMT Polyclonal Antibody for Western Blot, ICC/IF, IP

RNMT belongs to the mRNA cap methyltransferase family. RNMT is a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5'-cap structure of mRNAs. It binds RNA containing 5'-terminal GpppC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNMT
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Rnmt
Gene Accession No. O43148
Gene Alias 2610002P10Rik; AI848273; cm1p; CMT1; CMT1c; hcm1p; hCMT1; hMet; Kiaa0398; MET; mKIAA0398; mRNA (guanine-7-)methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; mRNA cap guanine-N7 methyltransferase; mRNA cap methyltransferase; RG7MT1; RNA (guanine-7-) methyltransferase; RNA guanine-7 methyltransferase; RNMT
Gene Symbols Rnmt
Host Species Rabbit
Immunogen E. coli-derived Human RNMT fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.