Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNMTL1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298513AF700
Description
RNMTL1 Polyclonal specifically detects RNMTL1 in Human samples. It is validated for Western Blot.Specifications
RNMTL1 | |
Polyclonal | |
Western Blot | |
EC 2.1.1, EC 2.1.1.-, FLJ10581, HC90, putative RNA methyltransferase, RNA methyltransferase like 1, RNA methyltransferase-like protein 1 | |
Produced in rabbits immunized with E. coli-derived Human RNMTL1 fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
55178 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction