Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNMTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554671 Supplier Invitrogen™ Supplier No. PA554671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILLEGRRLIS DALKAGAVPK MFFFSRLEYL KELPVDKLKG VSLIKVKFED IKDWSDLVTP QGIMGIFAKP DHVKMTYPKT QLQHSLPLLL ICDNLRDPGN Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 95%.

The RNA methyltransferase like 1 gene (RNMTL1) is one of thirteen newly discovered genes within a 116 Kb segment of the chromosome 17p13.3, which suffers from a high frequent loss of heterozygosity in human hepatocellular carcinoma. Its amino acid sequence was identified a high conserved region with methylases from various microorganisms, suggesting to have function to transfer methyl group to rRNA molecule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNMTL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRM3
Gene Accession No. Q9HC36
Gene Alias 16S rRNA (guanosine(1370)-2'-O)-methyltransferase; 16S rRNA [Gm1370] 2'-O-methyltransferase; 4833420N02Rik; AI255153; HC90; mitochondrial rRNA methyltransferase 3; MRM3; putative RNA methyltransferase; RGD1309077; RMTL1; RNA methyltransferase like 1; RNA methyltransferase-like protein 1; RNMTL1; rRNA methyltransferase 3, mitochondrial
Gene Symbols MRM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNMTL1. Recombinant protein control fragment (Product #RP-93453).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.