Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNPC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107249 Supplier Invitrogen™ Supplier No. PA5107249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 92%; Dog: 92%; Horse: 92%; Human: 100%; Mouse: 85%; Pig: 92%; Rabbit: 92%; Rat: 85%; Yeast: 77%.

Participates in pre-mRNA U12-dependent splicing, performed by the minor spliceosome which removes U12-type introns. U12-type introns comprises less than 1% of all non-coding sequences. Binds to the 3'-stem-loop of m(7)G-capped U12 snRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNPC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene RNPC3
Gene Accession No. Q96LT9
Gene Alias 2810441O16Rik; AI447568; C030014B17Rik; KIAA1839; Rbm40; RNA binding region (RNP1, RRM) containing 3; RNA recognition protein; RNA-binding motif protein 40; RNA-binding protein 40; RNA-binding region (RNP1, RRM) containing 3; RNA-binding region-containing protein 3; RNP; Rnpc3; SNRNP65; U11/U12 small nuclear ribonucleoprotein 65 kDa protein; U11/U12 snRNP 65 kDa protein; U11/U12 snRNP 65K; U11/U12-65K
Gene Symbols RNPC3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RNPC3 (aa 215-265).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55599
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.