Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNPEP Monoclonal Antibody (4E1)
GREENER_CHOICE

Catalog No. PIMA519830
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519830 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519830 Supplier Invitrogen™ Supplier No. MA519830
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: GNVKKLGDTY PSISNARNAE LRLRWGQIVL KNDHQEDFWK VKEFLHNQGK QKYTLPLYHA MMGGSEVAQT LAKETFASTA SQLHSNVVNY VQQIVAPKGS.

Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNPEP
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4E1
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Rnpep
Gene Accession No. Q9H4A4
Gene Alias aminopeptidase B; APB; AP-B; Arginine aminopeptidase; arginyl aminopeptidase; arginyl aminopeptidase (aminopeptidase B); cytosol aminopeptidase IV; Rnpep; zgc:100971
Gene Symbols Rnpep
Host Species Mouse
Immunogen RNPEP (NP_064601, 551 a.a. ~ 650 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6051
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.