Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNPEP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565165
Description
Immunogen sequence: MKPAEELAQLW AAEELDMKAI EAVAISPWKT YQLVYFLDKI LQKSPLPPGN VKKLGDTYPS ISNARNAELR LRWGQI Highest antigen sequence identity to the following orthologs - mouse 87%, rat 90%.
Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).
Specifications
| RNPEP | |
| Polyclonal | |
| Unconjugated | |
| Rnpep | |
| aminopeptidase B; APB; AP-B; Arginine aminopeptidase; arginyl aminopeptidase; arginyl aminopeptidase (aminopeptidase B); cytosol aminopeptidase IV; Rnpep; zgc:100971 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6051 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H4A4 | |
| Rnpep | |
| Recombinant Human RNPEP. Recombinant protein control fragment (Product #RP-104664). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction