Learn More
RNPEPL1 Antibody (11G1), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259373IR
Catalog No. NBP259373IR
Only null left
Add to Cart
Description
Description
RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.
Specifications
Specifications
RNPEPL1
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
RNPEPL1
Synthetic N-terminal arginylated peptide conjugated to KLH
100 μL
57140
Non-species specific
Purified
Western Blot
11G1
Western Blot
argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1
Mouse
Protein G purified
Primary
Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine.
Store at 4C in the dark.
IgG3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.