Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNPEPL1 Antibody (2A4), Novus Biologicals™

Mouse Monoclonal Antibody
$670.00
Specifications
Antigen | RNPEPL1 |
---|---|
Clone | 2A4 |
Dilution | Western Blot 1:100 - 1:2000 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.Specifications
RNPEPL1 | |
Western Blot 1:100 - 1:2000 | |
Unconjugated | |
Mouse | |
argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1 | |
RNPEPL1 | |
IgG3 | |
Protein G purified |
2A4 | |
Monoclonal | |
Purified | |
RUO | |
57140 | |
Synthetic N-terminal arginylated peptide conjugated to KLH | |
Primary | |
Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title