Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNPEPL1 Antibody (4D12), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259372AF488
Description
RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.Specifications
RNPEPL1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RNPEPL1 | |
Synthetic N-terminal arginylated peptide conjugated to KLH | |
100 μL | |
57140 | |
Non-species specific | |
Purified |
Western Blot | |
4D12 | |
Western Blot | |
argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1 | |
Mouse | |
Protein G purified | |
Primary | |
Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction