Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNPEPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140062
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LPQEVVMSLS KCYSSLLDSM NAEIRIRWLQ IVVRNDYYPD LHRVRRFLES QMSRMYTIPL YEDLCTGALK SFA.
RNPEPL1 has broad specificity aminopeptidase which preferentially hydrolyzes an N-terminal methionine, citrulline or glutamine..
Specifications
RNPEPL1 | |
Polyclonal | |
Unconjugated | |
RNPEPL1 | |
1110014H17Rik; AI894167; aminopeptidase B-like 1; Aminopeptidase RNPEPL1; argininyl aminopeptidase-like 1; arginyl aminopeptidase (aminopeptidase B)-like 1; arginyl aminopeptidase like 1; arginyl aminopeptidase-like 1; Methionyl aminopeptidase; RNPEPL1; RNPEP-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HAU8 | |
RNPEPL1 | |
Recombinant protein corresponding to Human RNPEPL1. Recombinant protein control fragment (Product #RP-108820). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction