Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560589 Supplier Invitrogen™ Supplier No. PA560589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKSLLGVKEN NKKSSTRAPS PTKRKDRSDE KSKDRSKDKG ATKESSEKDR GRDKTRKRRS ASSG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.

This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. This protein contains many serine residues. Two splice variants have been found for this gene; both variants encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNPS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNPS1
Gene Accession No. Q15287
Gene Alias E5.1; LDC2; ribonucleic acid binding protein S1; RNA binding protein S1; RNA binding protein S1, serine-rich domain; RNA binding protein with serine rich domain 1; RNA binding protein with serine-rich domain; RNA-binding protein S1, serine-rich domain; RNA-binding protein with serine-rich domain 1; Rnps1; SR protein; SR-related protein LDC2
Gene Symbols RNPS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNPS1. Recombinant protein control fragment (Product #RP-98811).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10921
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.