Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 15-4513
GREENER_CHOICE

Catalog No. 199710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-971-0 10 mg
19-975-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-971-0 Supplier Tocris Bioscience™ Supplier No. 1997/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Benzodiazepine partial inverse agonist

High affinity benzodiazepine ligand. Ki values are 3.1 and 5.3 nM for diazepam-insensitive (DI) and diazepam-sensitive (DS) benzodiazepine receptors respectively. Acts as partial inverse agonist at recombinant DS α1-, α2-, α3- and α5-GABAA receptors. Displays partial agonism at DI α4- and α6-GABAA receptors. Antagonizes several behavioral and neurochemical effects of ethanol. Proconvulsant and anxiogenic.

Chemical Identifiers

CAS 91917-65-6
Molecular Formula C15H14N6O3

Specifications

Quantity 10 mg
Formula Weight 326.31
Purity >98%
Molecular Weight (g/mol) 326.316
Chemical Name or Material Ro 15-4513
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.