Learn More
Tocris Bioscience™ Ro 25-6981 maleate
Description
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
| Chemical Name or Material | (αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate |
| CAS | 1312991-76-6 |
| Assay Percent Range | >98% |
| Quantity | 1 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C22H29NO2.C4H4O4 |
| Formula Weight | Observed MW: 455.55 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.