Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 32-0432 hydrochloride
GREENER_CHOICESDP

Catalog No. 15871 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-871 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-871 Supplier Tocris Bioscience™ Supplier No. 1587/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, orally active PKC inhibitor

Selective cell-permeable protein kinase C inhibitor. Displays slight selectivity for conventional PKC isoforms over Ca2+ and atypical PKC isoforms; binding affinities for rat isoforms are 9, 28, 31, 37 and 108 nM for PKC's α, βΙ, βΙΙ, γ and ε respectively. Orally available and prevents T cell chronic inflammation in vivo.

Chemical Identifiers

CAS 1781828-85-0
Molecular Formula C28H29ClN4O2

Specifications

Quantity 1 mg
Formula Weight 489.01
Purity >99%
Molecular Weight (g/mol) 489.016
Chemical Name or Material Ro 32-0432 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.